missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ALDH1 / ALDH1A1 / ALDC Protein

A DNA sequence encoding the Mus musculus (Mouse) ALDH1 / ALDH1A1 / ALDC, was expressed in the hosts and tags indicated.

2135.00 DKK - 29485.00 DKK

Specifications

Accession Number NP_038495.2
Gene ID (Entrez) 11668
Name ALDH1 / ALDH1A1 / ALDC Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16077321
View Documents
enQuireBio™
QP8967-ye-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16017331
View Documents
enQuireBio™
QP8967-ye-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16067321
View Documents
enQuireBio™
QP8967-ye-100ug
100 ÎĽg
6655.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16097321
View Documents
enQuireBio™
QP8967-ye-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16007331
View Documents
enQuireBio™
QP8967-ye-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16087321
View Documents
enQuireBio™
QP8967-ye-1mg
1 mg
29485.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_038495.2
ALDH1 / ALDH1A1 / ALDC Protein
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
11668
Research Use Only
Aldh1a1
Recombinant Protein
Yeast
SSPAQPAVPA PLADLKIQHT KIFINNEWHN SVSGKKFPVL NPATEEVICH VEEGDKADVD KAVKAARQAF QIGSPWRTMD ASERGRLLNK LADLMERDRL LLATMEALNG GKVFANAYLS DLGGCIKALK YCAGWADKIH GQTIPSDGDI FTYTRREPIG VCGQIIPWNF PMLMFIWKIG PALSCGNTVV VKPAEQTPLT ALHLASLIKE AGFPPGVVNI VPGYGPTAGA AISSHMDVDK VAFTGSTQVG KLIKEAAGKS NLKRVTLELG GKSPCIVFAD ADLDIAVEFA HHGVFYHQGQ CCVAASRIFV EESVYDEFVK RSVERAKKYV LGNPLTPGIN QGPQIDKEQH DKILDLIESG KKEGAKLECG GGRWGNKGFF VQPTVFSNVT DEMRIAKEEI FGPVQQIMKF KSVDDVIKRA NNTTYGLAAG LFTKDLDKAI TVSSALQAGV VWVNCYMMLS AQCPFGGFKM SGNGRELGEH GLYEYTELKT VAMKISQKNS
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse ALDH1 / ALDH1A1 / ALDC Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.