missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse ALDH1 / ALDH1A1 / ALDC Protein
A DNA sequence encoding the Mus musculus (Mouse) ALDH1 / ALDH1A1 / ALDC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8967-ye-200ug
This item is not returnable.
View return policy
Specifications
NP_038495.2 | |
ALDH1 / ALDH1A1 / ALDC Protein | |
Research Use Only | |
Aldh1a1 | |
Recombinant Protein | |
Yeast | |
SSPAQPAVPA PLADLKIQHT KIFINNEWHN SVSGKKFPVL NPATEEVICH VEEGDKADVD KAVKAARQAF QIGSPWRTMD ASERGRLLNK LADLMERDRL LLATMEALNG GKVFANAYLS DLGGCIKALK YCAGWADKIH GQTIPSDGDI FTYTRREPIG VCGQIIPWNF PMLMFIWKIG PALSCGNTVV VKPAEQTPLT ALHLASLIKE AGFPPGVVNI VPGYGPTAGA AISSHMDVDK VAFTGSTQVG KLIKEAAGKS NLKRVTLELG GKSPCIVFAD ADLDIAVEFA HHGVFYHQGQ CCVAASRIFV EESVYDEFVK RSVERAKKYV LGNPLTPGIN QGPQIDKEQH DKILDLIESG KKEGAKLECG GGRWGNKGFF VQPTVFSNVT DEMRIAKEEI FGPVQQIMKF KSVDDVIKRA NNTTYGLAAG LFTKDLDKAI TVSSALQAGV VWVNCYMMLS AQCPFGGFKM SGNGRELGEH GLYEYTELKT VAMKISQKNS | |
Greater than 90% as determined by SDS-PAGE. |
11668 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse ALDH1 / ALDH1A1 / ALDC Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction