missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse ALDH1 / ALDH1A1 / ALDC Protein

A DNA sequence encoding the Mus musculus (Mouse) ALDH1 / ALDH1A1 / ALDC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8967-ye-500ug

 View more versions of this product

Product Code. 16007331

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_038495.2
ALDH1 / ALDH1A1 / ALDC Protein
Research Use Only
Aldh1a1
Recombinant Protein
Yeast
SSPAQPAVPA PLADLKIQHT KIFINNEWHN SVSGKKFPVL NPATEEVICH VEEGDKADVD KAVKAARQAF QIGSPWRTMD ASERGRLLNK LADLMERDRL LLATMEALNG GKVFANAYLS DLGGCIKALK YCAGWADKIH GQTIPSDGDI FTYTRREPIG VCGQIIPWNF PMLMFIWKIG PALSCGNTVV VKPAEQTPLT ALHLASLIKE AGFPPGVVNI VPGYGPTAGA AISSHMDVDK VAFTGSTQVG KLIKEAAGKS NLKRVTLELG GKSPCIVFAD ADLDIAVEFA HHGVFYHQGQ CCVAASRIFV EESVYDEFVK RSVERAKKYV LGNPLTPGIN QGPQIDKEQH DKILDLIESG KKEGAKLECG GGRWGNKGFF VQPTVFSNVT DEMRIAKEEI FGPVQQIMKF KSVDDVIKRA NNTTYGLAAG LFTKDLDKAI TVSSALQAGV VWVNCYMMLS AQCPFGGFKM SGNGRELGEH GLYEYTELKT VAMKISQKNS
Greater than 90% as determined by SDS-PAGE.
11668
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Mouse ALDH1 / ALDH1A1 / ALDC Protein > 500μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.