missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uncharacterized protein C9orf163 Protein
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C9orf163, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_689784.1 |
---|---|
Gene ID (Entrez) | 158055 |
Name | Uncharacterized protein C9orf163 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16061191
|
enQuireBio™
QP7854-EC-100UG |
100 μg |
3450.00 DKK
100µg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081191
|
enQuireBio™
QP7854-EC-1MG |
1 mg |
15565.00 DKK
1mg |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071191
|
enQuireBio™
QP7854-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091191
|
enQuireBio™
QP7854-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001201
|
enQuireBio™
QP7854-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011201
|
enQuireBio™
QP7854-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_689784.1 | |
Uncharacterized protein C9orf163 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
158055 | |
Research Use Only | |
C9orf163 | |
Recombinant Protein | |
E. coli | |
MPGPLTCTPA WQGQGRAAAF LCCSFQRAGA VVGVPARWHR GRLSSQQRLR SSLGGSHPCP QLGRRLVREG VISVPRQQGR RRCRESFSPA DVAPGPICSA NICLSGVRFL TCLNRVREHV VGPSPSPAAP ICFFPVVEAL CTLRGRRCHC LPFPKRGMQR WMLPLRRGAR LLPLASSKNP RARSPGLDPL GSSETLWSHR GGH | |
Greater than 90% as determined by SDS-PAGE. |