missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Uncharacterized protein C9orf163 Protein
A DNA sequence encoding the Homo sapiens (Human) Uncharacterized protein C9orf163, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7854-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_689784.1 | |
Uncharacterized protein C9orf163 Protein | |
Research Use Only | |
C9orf163 | |
Recombinant Protein | |
E. coli | |
MPGPLTCTPA WQGQGRAAAF LCCSFQRAGA VVGVPARWHR GRLSSQQRLR SSLGGSHPCP QLGRRLVREG VISVPRQQGR RRCRESFSPA DVAPGPICSA NICLSGVRFL TCLNRVREHV VGPSPSPAAP ICFFPVVEAL CTLRGRRCHC LPFPKRGMQR WMLPLRRGAR LLPLASSKNP RARSPGLDPL GSSETLWSHR GGH | |
Greater than 90% as determined by SDS-PAGE. |
158055 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |