Learn More
enQuireBio™ Recombinant Human TSSK3 Protein
A DNA sequence encoding the Homo sapiens (Human) TSSK3, was expressed in the hosts and tags indicated.
1820.00 DKK - 22245.00 DKK
Specifications
Accession Number | NP_443073.1 |
---|---|
Gene ID (Entrez) | 81629 |
Name | TSSK3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16071871
|
enQuireBio™
QP7965-EC-100UG |
100 μg |
5015.00 DKK
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091871
|
enQuireBio™
QP7965-EC-1MG |
1 mg |
22245.00 DKK
1mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081871
|
enQuireBio™
QP7965-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001881
|
enQuireBio™
QP7965-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011881
|
enQuireBio™
QP7965-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021881
|
enQuireBio™
QP7965-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_443073.1 | |
TSSK3 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
81629 | |
Research Use Only | |
TSSK3 | |
Recombinant Protein | |
E. coli | |
MEDFLLSNGY QLGKTIGEGT YSKVKEAFSK KHQRKVAIKV IDKMGGPEEF IQRFLPRELQ IVRTLDHKNI IQVYEMLESA DGKICLVMEL AEGGDVFDCV LNGGPLPESR AKALFRQMVE AIRYCHGCGV AHRDLKCENA LLQGFNLKLT DFGFAKVLPK SHRELSQTFC GSTAYAAPEV LQGIPHDSKK GDVWSMGVVL YVMLCASLPF DDTDIPKMLW QQQKGVSFPT HLSISADCQD LLKRLLEPDM ILRPSIEEVS WHPWLAST | |
Greater than 80% as determined by SDS-PAGE. |