missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSSK3 Protein
A DNA sequence encoding the Homo sapiens (Human) TSSK3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7965-ec-200ug
Weitere Details : Weight : 0.01000kg
Spezifikation
NP_443073.1 | |
TSSK3 Protein | |
Research Use Only | |
TSSK3 | |
Recombinant Protein | |
E. coli | |
MEDFLLSNGY QLGKTIGEGT YSKVKEAFSK KHQRKVAIKV IDKMGGPEEF IQRFLPRELQ IVRTLDHKNI IQVYEMLESA DGKICLVMEL AEGGDVFDCV LNGGPLPESR AKALFRQMVE AIRYCHGCGV AHRDLKCENA LLQGFNLKLT DFGFAKVLPK SHRELSQTFC GSTAYAAPEV LQGIPHDSKK GDVWSMGVVL YVMLCASLPF DDTDIPKMLW QQQKGVSFPT HLSISADCQD LLKRLLEPDM ILRPSIEEVS WHPWLAST | |
Greater than 80% as determined by SDS-PAGE. |
81629 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |