missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human TNF receptor-associated factor 6 Protein Product Code.: 16006200

enQuireBio™ Recombinant Human TNF receptor-associated factor 6 Protein

Product Code. 16006200
200 μg, 200µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16006200

Brand: enQuireBio™ QP6838ec200ug

Quantity Price
1 5005.00 DKK / 200µg
5 4680.18 DKK / 200µg

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.

View alternative products
This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) TNF receptor-associated factor 6, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_004611.1
Gene ID (Entrez) 7189
Name TNF receptor-associated factor 6 Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol TRAF6
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MSLLNCENSC GSSQSESDCC VAMASSCSAV TKDDSVGGTA STGNLSSSFM EEIQGYDVEF DPPLESKYEC PICLMALREA VQTPCGHRFC KACIIKSIRD AGHKCPVDNE ILLENQLFPD NFAKREILSL MVKCPNEGCL HKMELRHLED HQAHCEFALM DCPQCQRPFQ KFHINIHILK DCPRRQVSCD NCAASMAFED KEIHDQNCPL ANVICEYCNT ILIREQMPNH YDLDCPTAPI PCTFSTFGCH EKMQRNHLAR HLQENTQSHM RMLAQAVHSL SVIPDSGYIS EVRNFQETIH QLEGRLVRQD HQIRELTAKM ETQSMYVSEL KRTIRTLEDK VAEIEAQQCN GIYIWKIGNF GMHLKCQEEE KPVVIHSPGF YTGKPGYKLC MRLHLQLPTA QRCANYISLF VHTMQGEYDS HLPWPFQGTI RLTILDQSEA PVRQNHEEIM DAKPELLAFQ RPTIPRNPKG FGYVTFMHLE ALRQRTFIKD DTLLVRCEVS TRFDMGSLRR EGFQPRSTDA GV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human TNF receptor-associated factor 6 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.