missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sperm-associated antigen 16 Protein
A DNA sequence encoding the Homo sapiens (Human) Sperm-associated antigen 16 protein, was expressed in the hosts and tags indicated.
1820.00 DKK - 22245.00 DKK
Specifications
Accession Number | NP_001020607.1 |
---|---|
Gene ID (Entrez) | 79582 |
Name | Sperm-associated antigen 16 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16071001
|
enQuireBio™
QP7822-EC-100UG |
100 μg |
5015.00 DKK
100µg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091001
|
enQuireBio™
QP7822-EC-1MG |
1 mg |
22245.00 DKK
1mg |
Estimated Shipment: 08-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16081001
|
enQuireBio™
QP7822-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001011
|
enQuireBio™
QP7822-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16011011
|
enQuireBio™
QP7822-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021011
|
enQuireBio™
QP7822-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001020607.1 | |
Sperm-associated antigen 16 Protein | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
79582 | |
Research Use Only | |
SPAG16 | |
Recombinant Protein | |
E. coli | |
MAAQRGMPSS AVRVLEEALG MGLTAAGDAR DTADAVAAEG AYYLEQVTIT EASEDDYEYE EIPDDNFSIP EGEEDLAKAI QMAQEQATDT EILERKTVLP SKHAVPEVIE DFLCNFLIKM GMTRTLDCFQ SEWYELIQKG VTELRTVGNV PDVYTQIMLL ENENKNLKKD LKHYKQAAEY VIF | |
Greater than 80% as determined by SDS-PAGE. |