missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sperm-associated antigen 16 Protein
A DNA sequence encoding the Homo sapiens (Human) Sperm-associated antigen 16 protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7822-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001020607.1 | |
Sperm-associated antigen 16 Protein | |
Research Use Only | |
SPAG16 | |
Recombinant Protein | |
E. coli | |
MAAQRGMPSS AVRVLEEALG MGLTAAGDAR DTADAVAAEG AYYLEQVTIT EASEDDYEYE EIPDDNFSIP EGEEDLAKAI QMAQEQATDT EILERKTVLP SKHAVPEVIE DFLCNFLIKM GMTRTLDCFQ SEWYELIQKG VTELRTVGNV PDVYTQIMLL ENENKNLKKD LKHYKQAAEY VIF | |
Greater than 80% as determined by SDS-PAGE. |
79582 | |
50 μg | |
Not determined. | |
Full Length of Isoform 4 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |