missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human Laminin subunit beta-2 Protein Product Code.: 16026471

enQuireBio™ Recombinant Human Laminin subunit beta-2 Protein

Product Code. 16026471
500 μg, 500µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16026471

Brand: enQuireBio™ QP8788ec500ug

Quantity Price
1 8750.00 DKK / 500µg
3 8182.13 DKK / 500µg

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.

View alternative products
This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Laminin subunit beta-2, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_002283.3
Gene ID (Entrez) 3913
Name Laminin subunit beta-2 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol LAMB2
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence PDSIEMVATR VLELSIPASA EQIQHLAGAI AERVRSLADV DAILARTVGD VRRAEQLLQD ARRARSWAED EKQKAETVQA ALEEAQRAQG IAQGAIRGAV ADTRDTEQTL YQVQERMAGA ERALSSAGER ARQLDALLEA LKLKRAGNSL AASTAEETAG SAQGRAQEAE QLLRGPLGDQ YQTVKALAER KAQGVLAAQA RAEQLRDEAR DLLQAAQDKL QRLQELEGTY EENERALESK AAQLDGLEAR MRSVLQAINL QVQIYNTCQ
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Laminin subunit beta-2 Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.