missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Guanine nucleotide-binding protein G(k) subunit alpha Protein
A DNA sequence encoding the Homo sapiens (Human) Guanine nucleotide-binding protein G(k) subunit alpha, was expressed in the hosts and tags indicated.
1185.00 DKK - 17250.00 DKK
Specifications
Accession Number | NP_006487.1 |
---|---|
Gene ID (Entrez) | 2773 |
Name | Guanine nucleotide-binding protein G(k) subunit alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16080800
|
enQuireBio™
QP6101-ec-10ug |
10 ÎĽg | N/A | ||||||
16020810
|
enQuireBio™
QP6101-ec-50ug |
50 ÎĽg | N/A | ||||||
16070800
|
enQuireBio™
QP6101-ec-100ug |
100 ÎĽg |
3825.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16000810
|
enQuireBio™
QP6101-ec-200ug |
200 ÎĽg | N/A | ||||||
16010810
|
enQuireBio™
QP6101-ec-500ug |
500 ÎĽg | N/A | ||||||
16090800
|
enQuireBio™
QP6101-ec-1mg |
1 mg |
17250.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_006487.1 | |
Guanine nucleotide-binding protein G(k) subunit alpha Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2773 | |
Research Use Only | |
GNAI3 | |
Recombinant Protein | |
E. coli | |
MGCTLSAEDK AAVERSKMID RNLREDGEKA AKEVKLLLLG AGESGKSTIV KQMKIIHEDG YSEDECKQYK VVVYSNTIQS IIAIIRAMGR LKIDFGEAAR ADDARQLFVL AGSAEEGVMT PELAGVIKRL WRDGGVQACF SRSREYQLND SASYYLNDLD RISQSNYIPT QQDVLRTRVK TTGIVETHFT FKDLYFKMFD VGGQRSERKK WIHCFEGVTA IIFCVALSDY DLVLAEDEEM NRMHESMKLF DSICNNKWFT ETSIILFLNK KDLFEEKIKR SPLTICYPEY TGSNTYEEAA AYIQCQFEDL NRRKDTKEIY THFTCATDTK NVQFVFDAVT DVIIKNNLKE CGLY | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Guanine nucleotide-binding protein G(k) subunit alpha Protein