missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Guanine nucleotide-binding protein G(k) subunit alpha Protein
A DNA sequence encoding the Homo sapiens (Human) Guanine nucleotide-binding protein G(k) subunit alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6101-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_006487.1 | |
Guanine nucleotide-binding protein G(k) subunit alpha Protein | |
Research Use Only | |
GNAI3 | |
Recombinant Protein | |
E. coli | |
MGCTLSAEDK AAVERSKMID RNLREDGEKA AKEVKLLLLG AGESGKSTIV KQMKIIHEDG YSEDECKQYK VVVYSNTIQS IIAIIRAMGR LKIDFGEAAR ADDARQLFVL AGSAEEGVMT PELAGVIKRL WRDGGVQACF SRSREYQLND SASYYLNDLD RISQSNYIPT QQDVLRTRVK TTGIVETHFT FKDLYFKMFD VGGQRSERKK WIHCFEGVTA IIFCVALSDY DLVLAEDEEM NRMHESMKLF DSICNNKWFT ETSIILFLNK KDLFEEKIKR SPLTICYPEY TGSNTYEEAA AYIQCQFEDL NRRKDTKEIY THFTCATDTK NVQFVFDAVT DVIIKNNLKE CGLY | |
Greater than 90% as determined by SDS-PAGE. |
2773 | |
500 ÎĽg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Guanine nucleotide-binding protein G(k) subunit alpha Protein > 500μg
Spot an opportunity for improvement?Share a Content Correction