missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3M Protein
A DNA sequence encoding the Homo sapiens (Human) EIF3M, was expressed in the hosts and tags indicated.
1385.00 DKK - 13770.00 DKK
Specifications
Accession Number | NP_001294858.1 |
---|---|
Gene ID (Entrez) | 10480 |
Name | EIF3M Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16070531
|
enQuireBio™
QP7743-EC-10UG |
10 μg |
1385.00 DKK
10µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090531
|
enQuireBio™
QP7743-EC-200UG |
200 μg |
5675.00 DKK
200µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060531
|
enQuireBio™
QP7743-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16010541
|
enQuireBio™
QP7743-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000541
|
enQuireBio™
QP7743-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080531
|
enQuireBio™
QP7743-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001294858.1 | |
EIF3M Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10480 | |
Research Use Only | |
EIF3M | |
Recombinant Protein | |
E. coli | |
SVPAFIDISE EDQAAELRAY LKSKGAEISE ENSEGGLHVD LAQIIEACDV CLKEDDKDVE SVMNSVVSLL LILEPDKQEA LIESLCEKLV KFREGERPSL RLQLLSNLFH GMDKNTPVRY TVYCSLIKVA ASCGAIQYIP TELDQVRKWI SDWNLTTEKK HTLLRLLYEA LVDCKKSDAA SKVMVELLGS YTEDNASQAR VDAHRCIVRA LKDPNAFLFD HLLTLKPVKF LEGELIHDLL TIFVSAKLAS YVKFYQNNKD FIDSLGLLHE QNMAKMRLLT FMGMAVENKE ISFDTMQQEL QIGADDVEAF VIDAVRTKMV YCKIDQTQRK VVVSHSTHRT FGKQQWQQLY DTLNAWKQNL NKVKNSLLSL SDT | |
Greater than 90% as determined by SDS-PAGE. |