missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF3M Protein
A DNA sequence encoding the Homo sapiens (Human) EIF3M, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7743-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001294858.1 | |
EIF3M Protein | |
Research Use Only | |
EIF3M | |
Recombinant Protein | |
E. coli | |
SVPAFIDISE EDQAAELRAY LKSKGAEISE ENSEGGLHVD LAQIIEACDV CLKEDDKDVE SVMNSVVSLL LILEPDKQEA LIESLCEKLV KFREGERPSL RLQLLSNLFH GMDKNTPVRY TVYCSLIKVA ASCGAIQYIP TELDQVRKWI SDWNLTTEKK HTLLRLLYEA LVDCKKSDAA SKVMVELLGS YTEDNASQAR VDAHRCIVRA LKDPNAFLFD HLLTLKPVKF LEGELIHDLL TIFVSAKLAS YVKFYQNNKD FIDSLGLLHE QNMAKMRLLT FMGMAVENKE ISFDTMQQEL QIGADDVEAF VIDAVRTKMV YCKIDQTQRK VVVSHSTHRT FGKQQWQQLY DTLNAWKQNL NKVKNSLLSL SDT | |
Greater than 90% as determined by SDS-PAGE. |
10480 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |