missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AN1-type zinc finger protein 1
A DNA sequence encoding the Homo sapiens (Human) AN1-type zinc finger protein 3, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_068762.1 |
---|---|
Gene ID (Entrez) | 60685 |
Name | AN1-type zinc finger protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16062601
|
enQuireBio™
QP8089-EC-100UG |
100 μg |
3450.00 DKK
100µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082601
|
enQuireBio™
QP8089-EC-1MG |
1 mg |
15565.00 DKK
1mg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16072601
|
enQuireBio™
QP8089-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092601
|
enQuireBio™
QP8089-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002611
|
enQuireBio™
QP8089-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012611
|
enQuireBio™
QP8089-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_068762.1 | |
AN1-type zinc finger protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
60685 | |
Research Use Only | |
ZFAND3 | |
Recombinant Protein | |
E. coli | |
MGDAGSERSK APSLPPRCPC GFWGSSKTMN LCSKCFADFQ KKQPDDDSAP STSNSQSDLF SEETTSDNNN TSITTPTLSP SQQPLPTELN VTSPSKEECG PCTDTAHVSL ITPTKRSCGT DSQSENEASP VKRPRLLENT ERSEETSRSK QKSRRRCFQC QTKLELVQQE LGSCRCGYVF CMLHRLPEQH DCTFDHMGRG REEAIMKMVK LDRKVGRSCQ RIGEGCS | |
Greater than 80% as determined by SDS-PAGE. |