missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AN1-type zinc finger protein 1
A DNA sequence encoding the Homo sapiens (Human) AN1-type zinc finger protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8089-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_068762.1 | |
AN1-type zinc finger protein 1 | |
Research Use Only | |
ZFAND3 | |
Recombinant Protein | |
E. coli | |
MGDAGSERSK APSLPPRCPC GFWGSSKTMN LCSKCFADFQ KKQPDDDSAP STSNSQSDLF SEETTSDNNN TSITTPTLSP SQQPLPTELN VTSPSKEECG PCTDTAHVSL ITPTKRSCGT DSQSENEASP VKRPRLLENT ERSEETSRSK QKSRRRCFQC QTKLELVQQE LGSCRCGYVF CMLHRLPEQH DCTFDHMGRG REEAIMKMVK LDRKVGRSCQ RIGEGCS | |
Greater than 80% as determined by SDS-PAGE. |
60685 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |