Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L30 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L30, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_000980.1 |
---|---|
Gene ID (Entrez) | 6156 |
Name | 60S ribosomal protein L30 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044600
|
enQuireBio™
QP6620-EC-100UG |
100 μg |
3450.00 DKK
100µg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064600
|
enQuireBio™
QP6620-EC-1MG |
1 mg |
15565.00 DKK
1mg |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054600
|
enQuireBio™
QP6620-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074600
|
enQuireBio™
QP6620-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084600
|
enQuireBio™
QP6620-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094600
|
enQuireBio™
QP6620-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000980.1 | |
60S ribosomal protein L30 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6156 | |
Research Use Only | |
RPL30 | |
Recombinant Protein | |
E. coli | |
MVAAKKTKKS LESINSRLQL VMKSGKYVLG YKQTLKMIRQ GKAKLVILAN NCPALRKSEI EYYAMLAKTG VHHYSGNNIE LGTACGKYYR VCTLAIIDPG DSDIIRSMPE QTGEK | |
Greater than 90% as determined by SDS-PAGE. |