missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L30 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L30, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6620-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000980.1 | |
60S ribosomal protein L30 Protein | |
Research Use Only | |
RPL30 | |
Recombinant Protein | |
E. coli | |
MVAAKKTKKS LESINSRLQL VMKSGKYVLG YKQTLKMIRQ GKAKLVILAN NCPALRKSEI EYYAMLAKTG VHHYSGNNIE LGTACGKYYR VCTLAIIDPG DSDIIRSMPE QTGEK | |
Greater than 90% as determined by SDS-PAGE. |
6156 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |