Learn More
enQuireBio™ Recombinant hepatitis Protein ORF3 Protein
A DNA sequence encoding the Hepatitis E virus genotype 1 (isolate Human/Pakistan/Sar-55) (HEV-1) Protein ORF3, was expressed in the hosts and tags indicated.
2300.00 DKK - 22245.00 DKK
Specifications
Name | hepatitis Protein ORF3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | ORF3 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068980
|
enQuireBio™
QP7492-EC-100UG |
100 μg |
6415.00 DKK
100µg |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088980
|
enQuireBio™
QP7492-EC-1MG |
1 mg |
22245.00 DKK
1mg |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078980
|
enQuireBio™
QP7492-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098980
|
enQuireBio™
QP7492-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008990
|
enQuireBio™
QP7492-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018990
|
enQuireBio™
QP7492-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
hepatitis Protein ORF3 Protein | |
Not determined. | |
Full Length | |
Hepatitis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
ORF3 | |
Recombinant Protein | |
E. coli | |
MGSRPWALGL FCCCSSCFCL CCSRHRPVSR LAAVVGGAAA VPAVVSGVTG LILSPSQSPI FIQPTPSPRM SPLRPGLDLV FANPSDHSAP LGATRPSAPP LPHVVDLPQL GPRR | |
Greater than 90% as determined by SDS-PAGE. |