missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant hepatitis Protein ORF3 Protein
A DNA sequence encoding the Hepatitis E virus genotype 1 (isolate Human/Pakistan/Sar-55) (HEV-1) Protein ORF3, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7492-ec-10ug
Additional Details : Weight : 0.01000kg
Spezifikation
hepatitis Protein ORF3 Protein | |
Research Use Only | |
ORF3 | |
Recombinant Protein | |
E. coli | |
MGSRPWALGL FCCCSSCFCL CCSRHRPVSR LAAVVGGAAA VPAVVSGVTG LILSPSQSPI FIQPTPSPRM SPLRPGLDLV FANPSDHSAP LGATRPSAPP LPHVVDLPQL GPRR | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Hepatitis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |