missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ribose-5-phosphate isomerase A, was expressed in the hosts and tags indicated.
3970.00 DKK - 31125.00 DKK
Specifications
Accession Number | NP_417389.1 |
---|---|
Gene ID (Entrez) | 947407 |
Name | Ribose-5-phosphate isomerase A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079070
|
enQuireBio™
QP7507-ye-10ug |
10 ÎĽg | N/A | ||||||
16019080
|
enQuireBio™
QP7507-ye-50ug |
50 ÎĽg | N/A | ||||||
16069070
|
enQuireBio™
QP7507-ye-100ug |
100 ÎĽg |
9855.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16099070
|
enQuireBio™
QP7507-ye-200ug |
200 ÎĽg | N/A | ||||||
16009080
|
enQuireBio™
QP7507-ye-500ug |
500 ÎĽg | N/A | ||||||
16089070
|
enQuireBio™
QP7507-ye-1mg |
1 mg |
31125.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_417389.1 | |
Ribose-5-phosphate isomerase A Protein | |
Not determined. | |
Full Length | |
E. coli | |
Untagged | |
Tris-based buffer, 50% glycerol |
947407 | |
Research Use Only | |
rpiA | |
Recombinant Protein | |
Yeast | |
MTQDELKKAV GWAALQYVQP GTIVGVGTGS TAAHFIDALG TMKGQIEGAV SSSDASTEKL KSLGIHVFDL NEVDSLGIYV DGADEINGHM QMIKGGGAAL TREKIIASVA EKFICIADAS KQVDILGKFP LPVEVIPMAR SAVARQLVKL GGRPEYRQGV VTDNGNVILD VHGMEILDPI AMENAINAIP GVVTVGLFAN RGADVALIGT PDGVKTIVK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein