Learn More
enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ribose-5-phosphate isomerase A, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7507-ye-10ug
 View more versions of this product
Product Code. 16079070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
NP_417389.1
Ribose-5-phosphate isomerase A Protein
Research Use Only
rpiA
Recombinant Protein
Yeast
MTQDELKKAV GWAALQYVQP GTIVGVGTGS TAAHFIDALG TMKGQIEGAV SSSDASTEKL KSLGIHVFDL NEVDSLGIYV DGADEINGHM QMIKGGGAAL TREKIIASVA EKFICIADAS KQVDILGKFP LPVEVIPMAR SAVARQLVKL GGRPEYRQGV VTDNGNVILD VHGMEILDPI AMENAINAIP GVVTVGLFAN RGADVALIGT PDGVKTIVK
Greater than 90% as determined by SDS-PAGE.
947407
10 μg
Not determined.
Full Length
E. coli
Untagged
Tris-based buffer, 50% glycerol
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein > 10μg