missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ribose-5-phosphate isomerase A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7507-ye-50ug
This item is not returnable.
View return policy
Specifications
NP_417389.1 | |
Ribose-5-phosphate isomerase A Protein | |
Research Use Only | |
rpiA | |
Recombinant Protein | |
Yeast | |
MTQDELKKAV GWAALQYVQP GTIVGVGTGS TAAHFIDALG TMKGQIEGAV SSSDASTEKL KSLGIHVFDL NEVDSLGIYV DGADEINGHM QMIKGGGAAL TREKIIASVA EKFICIADAS KQVDILGKFP LPVEVIPMAR SAVARQLVKL GGRPEYRQGV VTDNGNVILD VHGMEILDPI AMENAINAIP GVVTVGLFAN RGADVALIGT PDGVKTIVK | |
Greater than 90% as determined by SDS-PAGE. |
947407 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
Untagged | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Ribose-5-phosphate isomerase A Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction