missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Periplasmic serine endoprotease DegP Protein
A DNA sequence encoding the Escherichia coli (strain K12) Periplasmic serine endoprotease DegP, was expressed in the hosts and tags indicated.
2300.00 DKK - 24645.00 DKK
Specifications
Accession Number | NP_414703.1 |
---|---|
Gene ID (Entrez) | 947139 |
Name | Periplasmic serine endoprotease DegP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057010
|
enQuireBio™
QP7007-ec-10ug |
10 ÎĽg | N/A | ||||||
16097010
|
enQuireBio™
QP7007-ec-50ug |
50 ÎĽg | N/A | ||||||
16047010
|
enQuireBio™
QP7007-ec-100ug |
100 ÎĽg |
7110.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16077010
|
enQuireBio™
QP7007-ec-200ug |
200 ÎĽg | N/A | ||||||
16087010
|
enQuireBio™
QP7007-ec-500ug |
500 ÎĽg | N/A | ||||||
16067010
|
enQuireBio™
QP7007-ec-1mg |
1 mg |
24645.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_414703.1 | |
Periplasmic serine endoprotease DegP Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
947139 | |
Research Use Only | |
degP | |
Recombinant Protein | |
E. coli | |
AETSSATTAQ QMPSLAPMLE KVMPSVVSIN VEGSTTVNTP RMPRNFQQFF GDDSPFCQEG SPFQSSPFCQ GGQGGNGGGQ QQKFMALGSG VIIDADKGYV VTNNHVVDNA TVIKVQLSDG RKFDAKMVGK DPRSDIALIQ IQNPKNLTAI KMADSDALRV GDYTVAIGNP FGLGETVTSG IVSALGRSGL NAENYENFIQ TDAAINRGNS GGALVNLNGE LIGINTAILA PDGGNIGIGF AIPSNMVKNL TSQMVEYGQV KRGELGIMGT ELNSELAKAM KVDAQRGAFV SQVLPNSSAA KAGIKAGDVI TSLNGKPISS FAALRAQVGT MPVGSKLTLG LLRDGKQVNV NLELQQSSQN QVDSSSIFNG IEGAEMSNKG KDQGVVVNNV KTGTPAAQIG LKKGDVIIGA NQQAVKNIAE LRKVLDSKPS VLALNIQRGD STIYLLMQ | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Periplasmic serine endoprotease DegP Protein