missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Periplasmic serine endoprotease DegP Protein

A DNA sequence encoding the Escherichia coli (strain K12) Periplasmic serine endoprotease DegP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7007-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16067010

  • 22245.00 DKK / 1mg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_414703.1
Periplasmic serine endoprotease DegP Protein
Research Use Only
degP
Recombinant Protein
E. coli
AETSSATTAQ QMPSLAPMLE KVMPSVVSIN VEGSTTVNTP RMPRNFQQFF GDDSPFCQEG SPFQSSPFCQ GGQGGNGGGQ QQKFMALGSG VIIDADKGYV VTNNHVVDNA TVIKVQLSDG RKFDAKMVGK DPRSDIALIQ IQNPKNLTAI KMADSDALRV GDYTVAIGNP FGLGETVTSG IVSALGRSGL NAENYENFIQ TDAAINRGNS GGALVNLNGE LIGINTAILA PDGGNIGIGF AIPSNMVKNL TSQMVEYGQV KRGELGIMGT ELNSELAKAM KVDAQRGAFV SQVLPNSSAA KAGIKAGDVI TSLNGKPISS FAALRAQVGT MPVGSKLTLG LLRDGKQVNV NLELQQSSQN QVDSSSIFNG IEGAEMSNKG KDQGVVVNNV KTGTPAAQIG LKKGDVIIGA NQQAVKNIAE LRKVLDSKPS VLALNIQRGD STIYLLMQ
Greater than 90% as determined by SDS-PAGE.
947139
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers