missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Elongation factor P Protein
A DNA sequence encoding the Escherichia coli (strain K12) Elongation factor P, was expressed in the hosts and tags indicated.
2615.00 DKK - 29485.00 DKK
Specifications
Accession Number | WP_000257278.1 |
---|---|
Name | Elongation factor P Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | efp |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16078631
|
enQuireBio™
QP9742-ye-10ug |
10 ÎĽg | N/A | ||||||
16018641
|
enQuireBio™
QP9742-ye-50ug |
50 ÎĽg | N/A | ||||||
16068631
|
enQuireBio™
QP9742-ye-100ug |
100 ÎĽg |
7930.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16098631
|
enQuireBio™
QP9742-ye-200ug |
200 ÎĽg | N/A | ||||||
16008641
|
enQuireBio™
QP9742-ye-500ug |
500 ÎĽg | N/A | ||||||
16088631
|
enQuireBio™
QP9742-ye-1mg |
1 mg |
29485.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
WP_000257278.1 | |
Research Use Only | |
efp | |
Recombinant Protein | |
Yeast | |
MATYYSNDFR AGLKIMLDGE PYAVEASEFV KPGKGQAFAR VKLRRLLTGT RVEKTFKSTD SAEGADVVDM NLTYLYNDGE FWHFMNNETF EQLSADAKAI GDNAKWLLDQ AECIVTLWNG QPISVTPPNF VELEIVDTDP GLKGDTAGTG GKPATLSTGA VVKVPLFVQI GEVIKVDTRS GEYVSRVK | |
Greater than 90% as determined by SDS-PAGE. |
Elongation factor P Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Elongation factor P Protein