missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Elongation factor P Protein
A DNA sequence encoding the Escherichia coli (strain K12) Elongation factor P, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9742-ye-200ug
This item is not returnable.
View return policy
Specifications
WP_000257278.1 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
Elongation factor P Protein | |
Research Use Only | |
efp | |
Recombinant Protein | |
Yeast | |
MATYYSNDFR AGLKIMLDGE PYAVEASEFV KPGKGQAFAR VKLRRLLTGT RVEKTFKSTD SAEGADVVDM NLTYLYNDGE FWHFMNNETF EQLSADAKAI GDNAKWLLDQ AECIVTLWNG QPISVTPPNF VELEIVDTDP GLKGDTAGTG GKPATLSTGA VVKVPLFVQI GEVIKVDTRS GEYVSRVK | |
Greater than 90% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Elongation factor P Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction