missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Cytolethal distending toxin subunit B Protein

A DNA sequence encoding the Escherichia coli Cytolethal distending toxin subunit B, was expressed in the hosts and tags indicated.

2300.00 DKK - 24645.00 DKK

Specifications

Name Cytolethal distending toxin subunit B Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol cdtB
Protein Length Full Length of Mature Protein
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16080021
View Documents
enQuireBio™
QP7656-ec-10ug
10 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16020031
View Documents
enQuireBio™
QP7656-ec-50ug
50 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16070021
View Documents
enQuireBio™
QP7656-ec-100ug
100 ÎĽg
7110.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
16000031
View Documents
enQuireBio™
QP7656-ec-200ug
200 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16010031
View Documents
enQuireBio™
QP7656-ec-500ug
500 ÎĽg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16090021
View Documents
enQuireBio™
QP7656-ec-1mg
1 mg
24645.00 DKK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Cytolethal distending toxin subunit B Protein
Not determined.
Full Length of Mature Protein
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Research Use Only
cdtB
Recombinant Protein
E. coli
DLTDFRVATW NLQGASATTE SKWNINVRQL ISGENAVDIL AVQEAGSPPS TAVDTGTLIP SPGIPVRELI WNLSTNSRPQ QVYIYFSAVD ALGGRVNLAL VSNRRADEVF VLSPVRQGGR PLLGIRIGND AFFTAHAIAM RNNDAPALVE EVYNFFRDSR DPVHQALNWM ILGDFNREPA DLEMNLTVPV RRASEIISPA AATQTSQRTL DYAVAGNSVA FRPSPLQAGI VYGARRTQIS SDHFPVGVSR R
Greater than 80.40% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant E. coli Cytolethal distending toxin subunit B Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.