missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cytolethal distending toxin subunit B Protein
A DNA sequence encoding the Escherichia coli Cytolethal distending toxin subunit B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7656-ec-200ug
This item is not returnable.
View return policy
Specifications
Cytolethal distending toxin subunit B Protein | |
Research Use Only | |
cdtB | |
Recombinant Protein | |
E. coli | |
DLTDFRVATW NLQGASATTE SKWNINVRQL ISGENAVDIL AVQEAGSPPS TAVDTGTLIP SPGIPVRELI WNLSTNSRPQ QVYIYFSAVD ALGGRVNLAL VSNRRADEVF VLSPVRQGGR PLLGIRIGND AFFTAHAIAM RNNDAPALVE EVYNFFRDSR DPVHQALNWM ILGDFNREPA DLEMNLTVPV RRASEIISPA AATQTSQRTL DYAVAGNSVA FRPSPLQAGI VYGARRTQIS SDHFPVGVSR R | |
Greater than 80.40% as determined by SDS-PAGE. |
200 ÎĽg | |
Not determined. | |
Full Length of Mature Protein | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
CorrecciĂłn del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
TĂtulo del producto
enQuireBio™ Recombinant E. coli Cytolethal distending toxin subunit B Protein > 200μg
ÂżDetecta una oportunidad de mejora?Comparta una correcciĂłn de contenido