missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cytolethal distending toxin subunit B Protein
A DNA sequence encoding the Escherichia coli Cytolethal distending toxin subunit B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7656-ec-50ug
This item is not returnable.
View return policy
Specifications
Cytolethal distending toxin subunit B Protein | |
Research Use Only | |
cdtB | |
Recombinant Protein | |
E. coli | |
DLTDFRVATW NLQGASATTE SKWNINVRQL ISGENAVDIL AVQEAGSPPS TAVDTGTLIP SPGIPVRELI WNLSTNSRPQ QVYIYFSAVD ALGGRVNLAL VSNRRADEVF VLSPVRQGGR PLLGIRIGND AFFTAHAIAM RNNDAPALVE EVYNFFRDSR DPVHQALNWM ILGDFNREPA DLEMNLTVPV RRASEIISPA AATQTSQRTL DYAVAGNSVA FRPSPLQAGI VYGARRTQIS SDHFPVGVSR R | |
Greater than 80.40% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length of Mature Protein | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant E. coli Cytolethal distending toxin subunit B Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction