Learn More
Lenz Laborglasintrumente Reaction vessels, Cylindrical, with Thermostatic and Vacuum Jacket and Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
13535.00 DKK - 14550.00 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11903761
|
Lenz Laborglasintrumente
06061158 |
0.5 L |
14550.00 DKK
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
11983751
|
Lenz Laborglasintrumente
06061137 |
0.1 L |
13535.00 DKK
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
11993751
|
Lenz Laborglasintrumente
06061149 |
0.25 L |
13935.00 DKK
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
Description
- With graduation
- With laboratory flange LF
- With groove
- With two connectors (with ball flanges KF 15 /KF 25)
- Made of DURAN™ tubing
- Other sizes on request
Specifications
Duran Glass | |
1 Ea. |