Learn More
Lenz Laborglasintrumente Reaction vessels, Cylindrical, with Thermostatic and Vacuum Jacket and Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 06061137
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11983751
  • 13535.00 DKK / Each
Estimated Shipment: 16-08-2024
to see stock.
Add to basket
Description
Description
  • With graduation
  • With laboratory flange LF
  • With groove
  • With two connectors (with ball flanges KF 15 /KF 25)
  • Made of DURAN™ tubing
  • Other sizes on request
Specifications
Specifications
0.1 L
Duran Glass
280 mm
1 Ea.