missing translation for 'onlineSavingsMsg'
Få mere at vide

(R)-(-)-Deprenyl hydrochloride, Thermo Scientific™

Selective MAO-B inhibitor; anti-Parkinsonian agent

Ring for pris - 1820.92 DKK

Kemiske identifikatorer

CAS 14611-52-0
Molekylær formel C13H18ClN
Molekylvægt (g/mol) 223.74
MDL nummer MFCD00069299
InChI nøgle IYETZZCWLLUHIJ-UTONKHPSSA-N
Synonym selegiline hydrochloride, eldepryl, zelapar, selegiline hcl, l-deprenyl hydrochloride, jumex hydrochloride, --deprenyl hydrochloride, eldepryl hydrochloride, r-n-methyl-n-1-phenylpropan-2-yl prop-2-yn-1-amine hydrochloride, zydis selegiline
PubChem CID 26758
ChEBI CHEBI:9087
IUPAC navn hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
SMIL [H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C
Se flere specifikationer

Produkter 2
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
15483899
Dokumentvisning
Thermo Scientific Alfa Aesar
J61286.MD
250mg
Ring for pris
250mg
Se alternative produkter
Spar op til 
»
Ikke relevant
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

15473899
Dokumentvisning
Thermo Scientific Alfa Aesar
J61286.03
1g
1820.92 DKK
1g
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

Solubility
Soluble in water at 100mM

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Tekniske data

Kemiske identifikatorer

14611-52-0
223.74
IYETZZCWLLUHIJ-UTONKHPSSA-N
26758
hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
C13H18ClN
MFCD00069299
selegiline hydrochloride, eldepryl, zelapar, selegiline hcl, l-deprenyl hydrochloride, jumex hydrochloride, --deprenyl hydrochloride, eldepryl hydrochloride, r-n-methyl-n-1-phenylpropan-2-yl prop-2-yn-1-amine hydrochloride, zydis selegiline
CHEBI:9087
[H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C

Tekniske data

(R)-(-)-Deprenyl hydrochloride
C13H18ClN
14,8428
Soluble in water at 100mM
−12° (c=1 in Water)
hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
26758
223.74
14611-52-0
MFCD00069299
selegiline hydrochloride, eldepryl, zelapar, selegiline hcl, l-deprenyl hydrochloride, jumex hydrochloride, --deprenyl hydrochloride, eldepryl hydrochloride, r-n-methyl-n-1-phenylpropan-2-yl prop-2-yn-1-amine hydrochloride, zydis selegiline
IYETZZCWLLUHIJ-UTONKHPSSA-N
[H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C
223.74
CHEBI:9087
Videoer
Sikkerhed og håndtering

Sikkerhed og håndtering

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'rtecsNumber' : DA0292500

missing translation for 'tsca' : No

Anbefalet opbevaring : Ambient temperatures

SDS
Dokumenter

Dokumenter

Produktcertifikater

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.