missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ CentriVap™ Acid-Resistant Vacuum Concentration System

Offer maximum throughput processing upto 148 samples at once

Brand:  Labconco™ 7983014


Additional Details : CAS Number : 74-98-6 Weight : 50.00000kg

 View more versions of this product

Product Code. 12643105

  • 143425.00 DKK / Each
Estimated Shipment: 29-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Acid-resistant benchtop vacuum concentrator with glass lid (7810016), Cold Trap (7811020), Clear Canister (7460900), Acid Insert (7814800), 12-13mm Acid-Resistant Rotor (7397605), Glass Trap, Vacuum Tubing, 6 clamps

Requires : Diaphragm or rotary vane vacuum pump

Description

Description

Offer maximum throughput processing upto 148 samples at once

Vacuum Concentrator:

  • Acid-Resistant benchtop type with glass lid
  • Brushed stainless steel and glacier white, powder-coated steel exterior with blue accents
  • 300w heater with heat from OFF to HI (100°C) in one degree increments and heat time from 1 to 999 minutes
  • PTFE-coated chamber with Hastelloy shaft
Cold Trap:
  • HCFC/CFC-free refrigeration system with 1/3 hp compressor lowers cold trap to -50°C
  • Temperature graph that illuminates green when the collector coil reaches operating temperature

Certifications

CE marked (230V 50Hz model)

Compliance

ETL listed (115V model)

Specifications

Specifications

CE
Acid resistant concentrator with Glass lid, cold trap, clear canister, acid insert, 12 to13mm Acid resistant rotor, Glass trap and vacuum tubing and 6 clamp
Diaphragm or Rotary Vane
Benchtop
230 V
50 Hz
Glass
Schuko
148.0
230 V 50 Hz
4 A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions