Learn More
Kern Sohn Precision Balance EW
The classic balance with robust tuning fork measuring system.
Encompass_Preferred
5783.21 DKK - 8522.61 DKK
Specifications
Type Precision Balance
Electrical Requirements 230 V, 50 Hz
View More Specs
Products 7
Product Code Brand Capacity Price Quantity & Availability  
Product Code Brand Capacity Price Quantity & Availability  
17371603
Encompass_Preferred
View Documents
Kern Sohn
EW 12000-1NM
12 kg
6696.34 DKK
Each
View alternative products
Save Up to 
»
17391603
Encompass_Preferred
View Documents
Kern Sohn
EW 2200-2NM
2200 g
7076.81 DKK
Each
View alternative products
Save Up to 
»
17311613
Encompass_Preferred
View Documents
Kern Sohn
EW 4200-2NM
4200 g
7837.76 DKK
Each
View alternative products
Save Up to 
»
17301613
Encompass_Preferred
View Documents
Kern Sohn
EW 420-3NM
420 g
7381.19 DKK
Each
View alternative products
Save Up to 
»
17331613
Encompass_Preferred
View Documents
Kern Sohn
EW 6200-2NM
6200 g
8522.61 DKK
Each
View alternative products
Save Up to 
»
17321613
Encompass_Preferred
View Documents
Kern Sohn
EW 620-3NM
620 g
8218.23 DKK
Each
View alternative products
Save Up to 
»
17341613
Encompass_Preferred
View Documents
Kern Sohn
EW 820-2NM
820 g
5783.21 DKK
Each
View alternative products
Save Up to 
»
Description
Description
  • Adjusting program CAL for quick setting of the balance accuracy using an external test weight
  • Stable temperature behaviour
  • Short stabilisation time
  • Shock proof construction
  • High corner load performance
  • GLP/ISO record keeping of weight values
  • Totalising of pieces when counting
  • Protective working cover included with delivery
Specifications
Specifications
Precision Balance
230 V, 50 Hz
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn Precision Balance EW