missing translation for 'onlineSavingsMsg'
Learn More

Polaprezinc, MedChemExpress

MedChemExpress Polaprezinc is an orally bioavailable chelate composed of zinc and L-carnosine, with potential gastroprotective, anti-oxidant, anti-ulcer and anti-inflammatory activities.

Encompass_Preferred

261.85 DKK - 936.63 DKK

Specifications

Chemical Name or Material Polaprezinc
Physical Form Solid
CAS 107667-60-7
Color White
Purity Grade Notes Research
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16494457
Encompass_Preferred
View Documents
Medchem Express
HY-B0729
50 mg
624.42 DKK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16478588
Encompass_Preferred
View Documents
Medchem Express
HY-B0729
10 mg
261.85 DKK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16428436
Encompass_Preferred
View Documents
Medchem Express
HY-B0729
100 mg
936.63 DKK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Polaprezinc is an orally bioavailable chelate composed of zinc and L-carnosine, with potential gastroprotective, anti-oxidant, anti-ulcer and anti-inflammatory activities.
Specifications

Specifications

Polaprezinc
107667-60-7
Research
COVID-19-immunoregulation
H2O : 3.37 mg/mL (11.68 mM; ultrasonic and warming and adjust pH to 1-2 with HCl) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
288.59
98.0%
Solid
White
C9H11N4O3Zn
Zinc L-carnosine
O=C1[C@@H]([N-](C2=O)[Zn+2](NCC2)[O-]1)CC3=CN=CN3
288.59
Research
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Polaprezinc, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.