Learn More
Trajan Scientific and Medical™ Plunger Replacement For Fixed Needle Headspace Syringe
SGE syringe replacement plungers for gas tight syringes
359.00 DKK - 387.00 DKK
Specifications
Material PTFE (Plunger)
For Use With (Equipment) Thermo RSH Autosampler
Connection Type Gas Tight
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
13109195
View Documents Promotion Details
Trajan Scientific and Medical™
032841
1 mL
359.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 25-06-2024
to see stock.
13119195
View Documents Promotion Details
Trajan Scientific and Medical™
032846
2.5 mL
387.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 25-06-2024
to see stock.
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
  • For use with Thermo Scientific RSH Autosamplers
Specifications
Specifications
PTFE (Plunger)
Thermo RSH Autosampler
Gas Tight