Learn More
Trajan Scientific and Medical™ Plunger Replacement For Fixed Needle Headspace Syringe
SGE syringe replacement plungers for gas tight syringes
Brand:  Trajan Scientific and Medical™ 032846
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13119195
  • 387.00 DKK / Each
Estimated Shipment: 18-06-2024
to see stock.
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
  • For use with Thermo Scientific RSH Autosamplers
Specifications
Specifications
PTFE (Plunger)
2.5 mL
Thermo RSH Autosampler
Gas Tight