Learn More
Trajan Scientific and Medical™ Plunger Replacement For Fixed Needle Headspace Syringe
SGE syringe replacement plungers for gas tight syringes
Brand: Trajan Scientific and Medical™ 032841
Additional Details : Weight : 0.05000kg
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
- For use with Thermo Scientific RSH Autosamplers
Specifications
PTFE (Plunger) | |
1 mL | |
Thermo RSH Autosampler | |
Gas Tight |