Learn More
Pitolisant hydrochloride, MedChemExpress
MedChemExpress Pitolisant hydrochloride is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).

916.49 DKK - 2860.23 DKK
Specifications
Chemical Name or Material | Pitolisant hydrochloride |
---|---|
Physical Form | Solid |
CAS | 903576-44-3 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16425018
![]() |
Medchem Express
HY-12199B |
10 mg |
916.49 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16421257
![]() |
Medchem Express
HY-12199B |
50 mg |
2860.23 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pitolisant hydrochloride is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).Specifications
Pitolisant hydrochloride | |
903576-44-3 | |
Research | |
Neuroscience-Neuromodulation | |
H2O : 100 mg/mL (300.92 mM; Need ultrasonic) ∣DMSO : ≥ 43 mg/mL (129.40 mM) | |
332.31 | |
99.47% |
Solid | |
White | |
C17H27Cl2NO | |
Ciproxidine BF 2649 | |
ClC(C=C1)=CC=C1CCCOCCCN2CCCCC2.Cl | |
332.31 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.