missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pitolisant hydrochloride, MedChemExpress

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Pitolisant hydrochloride is a potent and selective nonimidazole inverse agonist at the recombinant human histamine H3 receptor (Ki=0.16 nM).
Specifications
Specifications
Chemical Name or Material | Pitolisant hydrochloride |
Physical Form | Solid |
CAS | 903576-44-3 |
Color | White |
Quantity | 50 mg |
Purity Grade Notes | Research |
Molecular Formula | C17H27Cl2NO |
For Use With (Application) | Neuroscience-Neuromodulation |
Synonym | Ciproxidine BF 2649 |
Solubility Information | H2O : 100 mg/mL (300.92 mM; Need ultrasonic) ∣DMSO : ≥ 43 mg/mL (129.40 mM) |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Pitolisant hydrochloride, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction