Learn More
BRAND™ Pipette Tray
Drawer organizer
189.00 DKK - 464.00 DKK
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
10261721
|
BRAND™
28300 |
Dimensions (L x W): 280 x 220 mm |
189.00 DKK
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
10684481
|
BRAND™
768515 |
Dimensions (L x W): 355 x 300 mm |
464.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
Description
Specifications
PVC |