Få mere at vide
Pipamperone Dihydrochloride, TRC
Artikelnummer. 30009758
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009758 100 mg
17961265 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009758 Leverandør Toronto Research Chemicals Leverandørnr. TRCP475200100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2448-68-2
Molekylær formel C21 H30 F N3 O2 . 2 Cl H
Molekylvægt (g/mol) 448.4
Synonym [1,4'-Bipiperidine]-4'-carboxamide, 1'-[3-(p-fluorobenzoyl)propyl]-, dihydrochloride, [1,4'-Bipiperidine]-4'-carboxamide, 1'-[4-(4-fluorophenyl)-4-oxobutyl]-, dihydrochloride, Pipamperone dichlorhydrate, Pipamperone dihydrochloride, Pipamperone hydrochloride, Piperonil, Propitan
IUPAC navn 1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidin-1-ylpiperidine-4-carboxamide;dihydrochloride
SMIL Cl.Cl.NC(=O)C1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)N3CCCCC3
Tekniske data
Kemisk navn eller materiale Pipamperone Dihydrochloride
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C21H30FN3O2.2ClH/c22-18-8-6-17(7-9-18)19(26)5-4-12-24-15-10-21(11-16-24,20(23)27)25-13-2-1-3-14-25;;/h6-9H,1-5,10-16H2,(H2,23,27);2*1H
Formel vægt 447.1856
Procent renhed >95%