CAS RN 2448-68-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Pipamperone Dihydrochloride, TRC
CAS: 2448-68-2 Molekylær formel: C21 H30 F N3 O2 . 2 Cl H Molekylvægt (g/mol): 448.4 Synonym: [1,4'-Bipiperidine]-4'-carboxamide, 1'-[3-(p-fluorobenzoyl)propyl]-, dihydrochloride,[1,4'-Bipiperidine]-4'-carboxamide, 1'-[4-(4-fluorophenyl)-4-oxobutyl]-, dihydrochloride,Pipamperone dichlorhydrate,Pipamperone dihydrochloride,Pipamperone hydrochloride,Piperonil,Propitan IUPAC navn: 1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidin-1-ylpiperidine-4-carboxamide;dihydrochloride SMIL: Cl.Cl.NC(=O)C1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)N3CCCCC3