Få mere at vide
Phenylbutazone, 98%, Thermo Scientific™
316.42 DKK - 805.98 DKK
Kemiske identifikatorer
| CAS | 50-33-9 |
|---|---|
| Molekylær formel | C19H20N2O2 |
| Molekylvægt (g/mol) | 308.38 |
| MDL nummer | MFCD00005500 |
| InChI nøgle | VYMDGNCVAMGZFE-UHFFFAOYSA-N |
| Synonym | phenylbutazone, fenilbutazona, butazolidin, diphenylbutazone, phenylbutazonum, butapirazol, butapyrazole, butazolidine, butacote, butadion |
| PubChem CID | 4781 |
| ChEBI | CHEBI:48574 |
| IUPAC navn | 4-butyl-1,2-diphenylpyrazolidine-3,5-dione |
| SMIL | CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1 |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
11302166
|
Thermo Scientific Alfa Aesar
L03449.14 |
25g |
316.42 DKK
25g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
11312166
|
Thermo Scientific Alfa Aesar
L03449.22 |
100g |
805.98 DKK
100g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| C19H20N2O2 | |
| MFCD00005500 | |
| phenylbutazone, fenilbutazona, butazolidin, diphenylbutazone, phenylbutazonum, butapirazol, butapyrazole, butazolidine, butacote, butadion | |
| CHEBI:48574 | |
| CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1 |
Tekniske data
| Phenylbutazone | |
| Odorless | |
| MFCD00005500 | |
| 290080 | |
| phenylbutazone, fenilbutazona, butazolidin, diphenylbutazone, phenylbutazonum, butapirazol, butapyrazole, butazolidine, butacote, butadion | |
| CCCCC1C(=O)N(N(C1=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 308.38 | |
| CHEBI:48574 | |
| 98% |
| 50-33-9 | |
| C19H20N2O2 | |
| UN2811 | |
| 14,7277 | |
| VYMDGNCVAMGZFE-UHFFFAOYSA-N | |
| 4-butyl-1,2-diphenylpyrazolidine-3,5-dione | |
| 4781 | |
| 308.38 |
Sikkerhed og håndtering
GHS H Statement
H301-H351-H361-H319
Toxic if swallowed.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes serious eye irritation.
GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : UQ8225000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.