missing translation for 'onlineSavingsMsg'
Learn More

pH 6.80 Buffer Solution at 20°C, Reagecon
BIOPHARMA

Tested, certified, NIST traceable, and shelf-stable buffer solution.

155.00 DKK - 227.00 DKK

Specifications

Description Accuracy ± 0.01 pH units
pH 6.80 ±0.01
Physical Form Liquid
Packaging HDPE Bottle GL45
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480459
BIOPHARMA
View Documents
Reagecon
10685
500 mL
155.00 DKK
500mL
Please to purchase this item. Need a web account? Register with us today!
11490459
BIOPHARMA
View Documents
Reagecon
1068
1 L
227.00 DKK
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications

Specifications

Accuracy ± 0.01 pH units
6.80 ±0.01
Liquid
HDPE Bottle GL45
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
pH 6.80 Buffer Solution at 20°C, Reagecon

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.