Learn More
pH 6.80 Buffer Solution at 20°C, Reagecon
Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand: Reagecon 10685
This item is not returnable.
View return policy
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Accuracy ± 0.01 pH units | |
Liquid | |
HDPE Bottle GL45 |
6.80 ±0.01 | |
500 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Product Title
pH 6.80 Buffer Solution at 20°C, Reagecon > 500 mL
Spot an opportunity for improvement?Share a Content Correction