missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 6.80 Buffer Solution at 20°C, Reagecon

Tested, certified, NIST traceable, and shelf-stable buffer solution.
Brand: Reagecon 1068
This item is not returnable.
View return policy
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Accuracy ± 0.01 pH units | |
Liquid | |
HDPE Bottle GL45 |
6.80 ±0.01 | |
1 L |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6822H1 | Certificate of Analysis | 20/09/2023 | 1068 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
pH 6.80 Buffer Solution at 20°C, Reagecon > 1 L
Spot an opportunity for improvement?Share a Content Correction