Learn More
Penicillin G potassium salt
Inhibits bacterial cell wall synthesis | CAS: 113-98-4 | C33H36ClKN4O4S2 | 691.34 g/mol
412.00 DKK - 1150.00 DKK
Chemical Identifiers
CAS | 113-98-4 |
---|---|
Molecular Formula | C33H36ClKN4O4S2 |
Molecular Weight (g/mol) | 691.34 |
MDL Number | MFCD00036193 |
InChI Key | HTQQVOYPSGNVPA-HBRCYENSSA-M |
Synonym | Benzylpenicillin potassium salt; Potassium benzylpenicillinate |
PubChem CID | 23664709 |
ChEBI | CHEBI:7963 |
IUPAC Name | potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylateShow More |
SMILES | [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O |
Description
Penicillin G potassium salt is used as a cell culture additive as an antibiotics. Use to inhibit the synthesis of bacterial cell walls by inhibition of the cell wall peptidoglycan chain cross-lining. It is the drug of choice for groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridians group streptococci, and non-penicillinase producing staphylococcus. The potassium salt has been used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPenicillin G potassium salt is used as a cell culture additive as an antibiotics. Use to inhibit the synthesis of bacterial cell walls by inhibition of the cell wall peptidoglycan chain cross-lining. It is the drug of choice for groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridians group streptococci, and non-penicillinase producing staphylococcus. The potassium salt has been used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.
Solubility
Soluble in water (100 mg/ml), methanol, ethanol (sparingly), and alcohol. Insoluble in chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
113-98-4 | |
691.34 | |
HTQQVOYPSGNVPA-HBRCYENSSA-M | |
23664709 | |
potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylateShow More |
C33H36ClKN4O4S2 | |
MFCD00036193 | |
Benzylpenicillin potassium salt; Potassium benzylpenicillinate | |
CHEBI:7963 | |
[K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=OShow More |
Specifications
Penicillin G Potassium Salt | |
113-98-4 | |
C33H36ClKN4O4S2 | |
3832841 | |
Benzylpenicillin potassium salt; Potassium benzylpenicillinate | |
HTQQVOYPSGNVPA-HBRCYENSSA-M | |
potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate | |
23664709 | |
372.48 |
Powder | |
White | |
MFCD00036193 | |
14,7094 | |
Soluble in water (100mg/ml),methanol,ethanol (sparingly),and alcohol. Insoluble in chloroform. | |
[K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O | |
691.34 | |
CHEBI:7963 | |
Penicillin G potassium salt |
Safety and Handling
GHS H Statement
H334-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
missing translation for 'einecsNumber' : 204-038-0
missing translation for 'rtecsNumber' : XH9700000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold